Learn More
BIRCH BIOTECH LLC PRISTINE Isopropyl Alcohol, HPLC Grade, 4 Liter, 4/case
Catalog No. NC3861155
Encompass_Preferred
PRISTINE Isopropyl Alcohol, HPLC Grade, 4 Liter, 4/caseHigh-Purity Solvent for High-Performance Liquid Chromatography (HPLC)PRISTINE HPLC Grade Isopropyl Alcohol is a high-purity solvent specially formulated for high-performance liquid chromatography and other demanding analytical applications. With a purity level of ≥99.9% and low levels of impurities, this solvent minimizes background noise, enabling accurate and reliable data collection. Ideal for applications requiring low UV absorbance and high consistency, it supports precise and interference-free results in both qualitative and quantitative analyses
Catalog No. NC3861155 Supplier BIRCH BIOTECH LLC Supplier No. 19145
PRISTINE Isopropyl Alcohol, HPLC Grade, 4 Liter, 4/caseHigh-Purity Solvent for High-Performance Liquid Chromatography (HPLC)PRISTINE HPLC Grade Isopropyl Alcohol is a high-purity solvent specially formulated for high-performance liquid chromatography and other demanding analytical applications. With a purity level of ≥99.9% and low levels of impurities, this solvent minimizes background noise, enabling accurate and reliable data collection. Ideal for applications requiring low UV absorbance and high consistency, it supports precise and interference-free results in both qualitative and quantitative analyses