Learn More
BIRCH BIOTECH LLC PRISTINE Isopropyl Alcohol, LC-MS Grade, 4 Liter, 4/case
Catalog No. NC3861154
Encompass_Preferred
PRISTINE Isopropyl Alcohol, LC-MS Grade, 4 Liter, 4/caseUltra-Pure Solvent for Liquid Chromatography-Mass Spectrometry (LC-MS)PRISTINE LC-MS Grade Isopropyl Alcohol is a premium, ultra-pure solvent specifically crafted for liquid chromatography-mass spectrometry and other high-sensitivity analytical techniques. With a purity level of ≥99.9% and minimal metal ions, this isopropyl alcohol is ideal for applications requiring maximum sensitivity and minimal background noise. Engineered for precision, it enables reliable, interference-free results in both qualitative and quantitative analyses.
Catalog No. NC3861154 Supplier BIRCH BIOTECH LLC Supplier No. 19345
PRISTINE Isopropyl Alcohol, LC-MS Grade, 4 Liter, 4/caseUltra-Pure Solvent for Liquid Chromatography-Mass Spectrometry (LC-MS)PRISTINE LC-MS Grade Isopropyl Alcohol is a premium, ultra-pure solvent specifically crafted for liquid chromatography-mass spectrometry and other high-sensitivity analytical techniques. With a purity level of ≥99.9% and minimal metal ions, this isopropyl alcohol is ideal for applications requiring maximum sensitivity and minimal background noise. Engineered for precision, it enables reliable, interference-free results in both qualitative and quantitative analyses.