Learn More
BIRCH BIOTECH LLC PRISTINE Methanol, HPLC Grade, 4 Liter, 4/case
Catalog No. NC3861145
Encompass_Preferred
PRISTINE Methanol, HPLC Grade, 4 Liter, 4/casePrecision Solvent for High-Performance Liquid Chromatography (HPLC)PRISTINE HPLC Grade Methanol is a high-purity solvent crafted for precision and reliability in high-performance liquid chromatography applications. With a purity level of ≥99.9%, it meets the stringent requirements for accurate, interference-free analysis in a variety of laboratory settings. This methanol grade is ideal for analytical methods requiring high sensitivity, such as UV detection, due to its low absorbance and consistent performance.
Catalog No. NC3861145 Supplier BIRCH BIOTECH LLC Supplier No. 19155
PRISTINE Methanol, HPLC Grade, 4 Liter, 4/casePrecision Solvent for High-Performance Liquid Chromatography (HPLC)PRISTINE HPLC Grade Methanol is a high-purity solvent crafted for precision and reliability in high-performance liquid chromatography applications. With a purity level of ≥99.9%, it meets the stringent requirements for accurate, interference-free analysis in a variety of laboratory settings. This methanol grade is ideal for analytical methods requiring high sensitivity, such as UV detection, due to its low absorbance and consistent performance.