Learn More
BIRCH BIOTECH LLC PRISTINE Methanol, LC-MS Grade, 4 Liter, 4/case
Catalog No. NC3861143
Encompass_Preferred
PRISTINE Methanol, LC-MS Grade, 4 Liter, 4/caseUltra-Pure Solvent for Liquid Chromatography-Mass Spectrometry (LC-MS)PRISTINE LC-MS Grade Methanol is a premium, ultra-pure solvent designed to meet the rigorous demands of liquid chromatography-mass spectrometry and other high-sensitivity analytical techniques. With a purity level of ≥99.9% and minimal levels of metal ions and organic contaminants, this methanol grade delivers consistent, high-performance results in both quantitative and qualitative analyses
Catalog No. NC3861143 Supplier BIRCH BIOTECH LLC Supplier No. 19355
PRISTINE Methanol, LC-MS Grade, 4 Liter, 4/caseUltra-Pure Solvent for Liquid Chromatography-Mass Spectrometry (LC-MS)PRISTINE LC-MS Grade Methanol is a premium, ultra-pure solvent designed to meet the rigorous demands of liquid chromatography-mass spectrometry and other high-sensitivity analytical techniques. With a purity level of ≥99.9% and minimal levels of metal ions and organic contaminants, this methanol grade delivers consistent, high-performance results in both quantitative and qualitative analyses