Learn More
BIRCH BIOTECH LLC PRISTINE Water, LC-MS Grade, 4 Liter, 4/case
Catalog No. NC3861146
Encompass_Preferred
PRISTINE Water, LC-MS Grade, 4 Liter, 4/caseUltra-Pure Water for Liquid Chromatography-Mass Spectrometry (LC-MS)PRISTINE LC-MS Grade Water is purified for the specific needs of liquid chromatography-mass spectrometry and other highly sensitive analytical techniques. With a purity level that minimizes ion suppression and interference, this water grade is designed for maximum sensitivity and accuracy in critical applications. Featuring ultra-low conductivity, low total organic carbon (TOC), and low UV absorbance, it is ideal for achieving reliable, precise results in both qualitative and quantitative analyses.
Catalog No. NC3861146 Supplier BIRCH BIOTECH LLC Supplier No. 19395
PRISTINE Water, LC-MS Grade, 4 Liter, 4/caseUltra-Pure Water for Liquid Chromatography-Mass Spectrometry (LC-MS)PRISTINE LC-MS Grade Water is purified for the specific needs of liquid chromatography-mass spectrometry and other highly sensitive analytical techniques. With a purity level that minimizes ion suppression and interference, this water grade is designed for maximum sensitivity and accuracy in critical applications. Featuring ultra-low conductivity, low total organic carbon (TOC), and low UV absorbance, it is ideal for achieving reliable, precise results in both qualitative and quantitative analyses.