Learn More
Medchemexpress LLC Pritelivir mesylate | 1428333-96-3 | 99.5% | 498.60 g·mol⁻¹ | C19H22N4O6S3 | 10 MG
SDP
Catalog No. 5000287337
Encompass_Preferred
Pritelivir mesylate is the mesylate salt of pritelivir, a small-molecule inhibitor of the herpes simplex virus helicase-primase complex. It demonstrates potent antiviral activity in vitro against HSV-1 and HSV-2 (reported IC50 ≈ 0.02 μM) and is intended for research use in antiviral and virology studies.
  • Inhibits the viral helicase-primase complex, reducing HSV replication.
  • Potent in vitro activity with low-nanomolar IC50.
  • High reported purity suitable for biochemical and cell-based assays.
  • Supplied as a mesylate salt for improved stability and solubility handling.
  • Available in small research quantities for preclinical experimentation.
Catalog No. 50-002-87337 Supplier Medchemexpress LLC Supplier No. HY15303A10MG
Pritelivir mesylate is the mesylate salt of pritelivir, a small-molecule inhibitor of the herpes simplex virus helicase-primase complex. It demonstrates potent antiviral activity in vitro against HSV-1 and HSV-2 (reported IC50 ≈ 0.02 μM) and is intended for research use in antiviral and virology studies.
  • Inhibits the viral helicase-primase complex, reducing HSV replication.
  • Potent in vitro activity with low-nanomolar IC50.
  • High reported purity suitable for biochemical and cell-based assays.
  • Supplied as a mesylate salt for improved stability and solubility handling.
  • Available in small research quantities for preclinical experimentation.