Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pritelivir mesylate | 1428333-96-3 | C19H22N4O6S3 | 100 MG
SDP

Catalog No. 5000373769
Encompass_Preferred

Pritelivir mesylate is an inhibitor of the viral helicase-primase complex. It demonstrates antiviral activity both in vitro and in animal models of herpes simplex virus (HSV) infection, effective against herpes simplex virus types 1 and 2 (HSV-1 and HSV-2).

  • Inhibits viral helicase-primase complex
  • Exhibits antiviral activity against HSV-1 and HSV-2
  • IC50 of 0.02 μM against HSV1-2
  • For research use only
  • Purity of 99.33%
  • Appearance is solid
  • Color is white to off-white

Catalog No. 50-003-73769 Supplier Medchemexpress LLC Supplier No. HY15303A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Pritelivir mesylate is an inhibitor of the viral helicase-primase complex. It demonstrates antiviral activity both in vitro and in animal models of herpes simplex virus (HSV) infection, effective against herpes simplex virus types 1 and 2 (HSV-1 and HSV-2).

  • Inhibits viral helicase-primase complex
  • Exhibits antiviral activity against HSV-1 and HSV-2
  • IC50 of 0.02 μM against HSV1-2
  • For research use only
  • Purity of 99.33%
  • Appearance is solid
  • Color is white to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.