Learn More
Medchemexpress LLC Pritelivir (mesylate) | 1428333-96-3 | C19H22N4O6S3 | 25 MG
SDP
Catalog No. 5000375389
Encompass_Preferred
Pritelivir mesylate is a potent inhibitor of the viral helicase-primase complex, demonstrating significant antiviral activity against herpes simplex virus types 1 and 2 (HSV-1 and HSV-2). It represents a novel class of antiviral agents with potential for treating severe HSV infections.
  • Inhibits viral helicase-primase complex
  • Shows antiviral activity against HSV-1 and HSV-2 with an IC50 of 0.02 μM
  • Increases survival in animal models of HSV infection
  • Exhibits resistance-breaking antiviral efficacy
  • Potential treatment for life-threatening HSV type 1 and 2 infections
Catalog No. 50-003-75389 Supplier Medchemexpress LLC Supplier No. 25MGHY15303A
Pritelivir mesylate is a potent inhibitor of the viral helicase-primase complex, demonstrating significant antiviral activity against herpes simplex virus types 1 and 2 (HSV-1 and HSV-2). It represents a novel class of antiviral agents with potential for treating severe HSV infections.
  • Inhibits viral helicase-primase complex
  • Shows antiviral activity against HSV-1 and HSV-2 with an IC50 of 0.02 μM
  • Increases survival in animal models of HSV infection
  • Exhibits resistance-breaking antiviral efficacy
  • Potential treatment for life-threatening HSV type 1 and 2 infections