Learn More
Medchemexpress LLC Pritelivir mesylate | 1428333-96-3 | C19H22N4O6S3 | 5 MG
SDP
Catalog No. 5000373895
Encompass_Preferred
Pritelivir mesylate is an inhibitor of the viral helicase-primase complex, demonstrating antiviral activity both in vitro and in animal models of herpes simplex virus (HSV) infection. It is effective against herpes simplex virus types 1 and 2 (HSV-1 and HSV-2) with an IC50 of 0.02 μM. As the first in its class of antiviral agents, it inhibits HSV replication by targeting the viral helicase-primase enzyme complex.
  • Inhibits the viral helicase-primase complex.
  • Active against herpes simplex virus types 1 and 2.
  • Effective with an IC50 of 0.02 μM against HSV1-2.
  • Significantly increases survival and reduces mortality against HSV-1 and HSV-2 in animal models.
  • Exhibits potent and resistance-breaking antiviral efficacy.
  • Potential treatment for life-threatening HSV infections.
Catalog No. 50-003-73895 Supplier Medchemexpress LLC Supplier No. HY15303A5MG
Pritelivir mesylate is an inhibitor of the viral helicase-primase complex, demonstrating antiviral activity both in vitro and in animal models of herpes simplex virus (HSV) infection. It is effective against herpes simplex virus types 1 and 2 (HSV-1 and HSV-2) with an IC50 of 0.02 μM. As the first in its class of antiviral agents, it inhibits HSV replication by targeting the viral helicase-primase enzyme complex.
  • Inhibits the viral helicase-primase complex.
  • Active against herpes simplex virus types 1 and 2.
  • Effective with an IC50 of 0.02 μM against HSV1-2.
  • Significantly increases survival and reduces mortality against HSV-1 and HSV-2 in animal models.
  • Exhibits potent and resistance-breaking antiviral efficacy.
  • Potential treatment for life-threatening HSV infections.