Learn More
Medchemexpress LLC Pritelivir mesylate | 1428333-96-3 | C19H22N4O6S3 | 50 MG
SDP
Catalog No. 5000372030
Encompass_Preferred
Pritelivir mesylate (BAY 57-1293 mesylate) is an inhibitor of the viral helicase-primase complex, demonstrating antiviral activity both in vitro and in animal models of herpes simplex virus (HSV) infection. It is active against HSV-1 and HSV-2 with an IC50 of 0.02 μM, making it the first in a class of antiviral agents to target this complex for HSV replication inhibition.
  • Exhibits antiviral activity against HSV-1 and HSV-2
  • Inhibits the viral helicase-primase complex
  • Significantly increases survival and reduces mortality in animal models of HSV-1 infection
  • Possesses potent and resistance-breaking antiviral efficacy
  • Potential for treating life-threatening HSV type 1 and 2 infections, including herpes simplex encephalitis
  • Protective when used in combination therapies with Acyclovir against HSV-2
Catalog No. 50-003-72030 Supplier Medchemexpress LLC Supplier No. HY15303A50MG
Pritelivir mesylate (BAY 57-1293 mesylate) is an inhibitor of the viral helicase-primase complex, demonstrating antiviral activity both in vitro and in animal models of herpes simplex virus (HSV) infection. It is active against HSV-1 and HSV-2 with an IC50 of 0.02 μM, making it the first in a class of antiviral agents to target this complex for HSV replication inhibition.
  • Exhibits antiviral activity against HSV-1 and HSV-2
  • Inhibits the viral helicase-primase complex
  • Significantly increases survival and reduces mortality in animal models of HSV-1 infection
  • Possesses potent and resistance-breaking antiviral efficacy
  • Potential for treating life-threatening HSV type 1 and 2 infections, including herpes simplex encephalitis
  • Protective when used in combination therapies with Acyclovir against HSV-2