Learn More
MSC PRIVO EYEWEAR
Catalog No. 5000782233
Encompass
Safety glasses from the Privo series are designed with clear polycarbonate lenses that feature both anti-fog and scratch-resistant coatings. They offer a half-framed, wrap-around design with a black frame and non-adjustable temples, suitable for universal fit.
  • Anti-fog coatings improve visibility under hot and humid conditions
  • Scratch-resistant lenses enhance durability
  • Half-framed design with wrap-around frame
  • Clear polycarbonate lens material
  • Non-adjustable temples with a soft nose bridge for comfort
  • UV protection included
  • Meets CSA Z94.3 and ANSI Z87.1-2015 standards
Catalog No. 50-007-82233 Supplier MSC Supplier No. 85684884
Safety glasses from the Privo series are designed with clear polycarbonate lenses that feature both anti-fog and scratch-resistant coatings. They offer a half-framed, wrap-around design with a black frame and non-adjustable temples, suitable for universal fit.
  • Anti-fog coatings improve visibility under hot and humid conditions
  • Scratch-resistant lenses enhance durability
  • Half-framed design with wrap-around frame
  • Clear polycarbonate lens material
  • Non-adjustable temples with a soft nose bridge for comfort
  • UV protection included
  • Meets CSA Z94.3 and ANSI Z87.1-2015 standards