Learn More
Sino Biological Recombinant Listeria monocytogenes PrKA Protein (full length, GST Tag)
Catalog No. 5000083950
Encompass_Preferred
Recombinant full-length Listeria monocytogenes serovar 1/2a PrkA was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-000-83950 Supplier Sino Biological Supplier No. P10134G20
Recombinant full-length Listeria monocytogenes serovar 1/2a PrkA was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.