Learn More
Aviva Systems Biology Corp Prkaa1 Antibody - N-terminal region (ARP53651_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53651_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Prkaa1. It was validated on Western Blot. . Alternate names: AI194361, AI450832, AL024255, AMPKalpha1, C130083N04RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-285-864