Learn More
Aviva Systems Biology Corp PRKAA1 Antibody - N-terminal region (ARP53846_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53846_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRKAA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PRKAA1. Alternate names: AMPK, AMPKa1, MGC33776, MGC57364ApplicationWB, IHC. Species reactivity: Human, Rabbit, Rat
Catalog No. 50-014-61146