Learn More
Aviva Systems Biology Corp PRKAA2 Antibody - middle region (ARP56697_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56697_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRKAA2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PRKAA2. Alternate names: AMPK, AMPK2, PRKAA, AMPKa2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-61957