Learn More
Aviva Systems Biology Corp PRKACA Antibody - N-terminal region (ARP57827_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57827_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRKACA. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PRKACA. Alternate names: MGC102831, MGC48865, PKACAApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-396