Learn More
Aviva Systems Biology Corp PRKACB Antibody - N-terminal region (ARP56415_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56415_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRKACB. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PRKACB. Alternate names: DKFZp781I2452, MGC41879, MGC9320, PKACBApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-150