Learn More
Sino Biological PRKAG1/PRKAB1/PRKAA1 Protein 50ug (Sino Biological)
Supplier:  Sino Biological CT008H0907B50
Encompass_Preferred
A DNA sequence encoding the human PRKAG1 (P54619) (Met 1-Pro 331), constructed the plasmid 1; A DNA sequence encoding the human PRKAB1 (O43741) (Met 1-Ile 272), constructed the plasmid 2; A DNA sequence encoding the human PRKAA2 (P54646) (Met 1-Arg 552) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus, constructed the plasmid 3. The three plasmids were co-expressed and the heterotrimer was purified.
Catalog No. 50-237-1725