Learn More
Aviva Systems Biology Corp PRKAR1A Antibody - C-terminal region (ARP57832_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57832_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRKAR1A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human PRKAR1A. Alternate names: CAR, CNC, CNC1, DKFZp779L0468, MGC17251, PKR1, PPNAD1, PRKAR1, TSE1, ADOHRApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-399