Learn More
Aviva Systems Biology Corp PRKAR1B Antibody - middle region (ARP56420_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56420_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRKAR1B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PRKAR1B. Alternate names: PRKAR1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-61853