Learn More
Aviva Systems Biology Corp PRKAR2A Antibody - middle region (ARP61563_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61563_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRKAR2A. It was validated on Western Blot. . Alternate names: MGC3606, PKR2, PRKAR2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-409