Learn More
Aviva Systems Biology Corp PRKCH Antibody - N-terminal region (ARP56703_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56703_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRKCH. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PRKCH. Alternate names: MGC26269, MGC5363, PKC-L, PKCL, PRKCL, nPKC-etaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-286-198