Learn More
Aviva Systems Biology Corp PRKCI Antibody - middle region (ARP56427_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56427_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRKCI. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human PRKCI. Alternate names: DXS1179E, MGC26534, PKCI, nPKC-iotaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-151