Learn More
Sino Biological Recombinant Human PRKD1/PKC mu Protein (Full Length, GST Tag), Active
Catalog No. 502573556
Encompass_Preferred
Recombinant full length human PKC mu was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.
Catalog No. 50-257-3556 Supplier Sino Biological Supplier No. P7210G500
Recombinant full length human PKC mu was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.