Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Parkin (PRKN) recombinant protein, N-SUMO, N-6xHis tagged | >95.0% | 100UG
SDP

Catalog No. 5000174361
Encompass_Preferred

Recombinant human Parkin (PRKN) protein, full-length isoform-1 (M1-V465), expressed in E. coli and provided as an N-terminal SUMO and 6xHis tagged, lyophilized preparation. The protein is >95% pure by reducing SDS-PAGE, has an approximate molecular weight of 70 kDa, and is formulated for stability in Tris-HCl/NaCl with TCEP, Brij-35, and glycerol. Storage and handling recommendations are provided by the manufacturer.

  • Full-length human isoform-1 (M1-V465)
  • N-terminal SUMO and 6xHis tags for affinity purification
  • Purity greater than 95% (reducing SDS-PAGE)
  • Approximate molecular weight ~70 kDa
  • Lyophilized formulation from filtered buffer for stability
  • Expression system: Escherichia coli
  • Recommended storage at -20°C or -80°C after aliquoting
  • Suitable for ubiquitination, mitophagy, and interaction assays

Catalog No. 50-001-74361 Supplier Medchemexpress LLC Supplier No. HYP702806100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Parkin (PRKN) protein, full-length isoform-1 (M1-V465), expressed in E. coli and provided as an N-terminal SUMO and 6xHis tagged, lyophilized preparation. The protein is >95% pure by reducing SDS-PAGE, has an approximate molecular weight of 70 kDa, and is formulated for stability in Tris-HCl/NaCl with TCEP, Brij-35, and glycerol. Storage and handling recommendations are provided by the manufacturer.

  • Full-length human isoform-1 (M1-V465)
  • N-terminal SUMO and 6xHis tags for affinity purification
  • Purity greater than 95% (reducing SDS-PAGE)
  • Approximate molecular weight ~70 kDa
  • Lyophilized formulation from filtered buffer for stability
  • Expression system: Escherichia coli
  • Recommended storage at -20°C or -80°C after aliquoting
  • Suitable for ubiquitination, mitophagy, and interaction assays

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.