Learn More
Medchemexpress LLC Recombinant human Parkin (PRKN) protein (His-SUMO tagged) | >95.0% | 10UG
SDP
Catalog No. 5000174171
Encompass_Preferred
Recombinant full-length human Parkin (PRKN) is expressed in E. coli and supplied with N-terminal SUMO and 6xHis affinity tags. The lyophilized protein is formulated for biochemical assays and antibody validation, with reported >95% purity and an apparent molecular weight near 70 kDa.
  • Full-length human sequence (M1-V465) suitable for functional and binding studies.
  • N-terminal SUMO and 6xHis tags for enhanced solubility and affinity purification.
  • Lyophilized formulation from Tris-HCl/NaCl with TCEP, Brij-35, and glycerol.
  • High purity (>95%) by reducing SDS-PAGE, minimizing assay interference.
  • Stable frozen; aliquot and store at -20°C or -80°C after reconstitution for long-term use.
  • Validated for antibody binding in immunoassays (reported ED50 ~79 ng/mL).
Catalog No. 50-001-74171 Supplier Medchemexpress LLC Supplier No. HYP70280610UG
Recombinant full-length human Parkin (PRKN) is expressed in E. coli and supplied with N-terminal SUMO and 6xHis affinity tags. The lyophilized protein is formulated for biochemical assays and antibody validation, with reported >95% purity and an apparent molecular weight near 70 kDa.
  • Full-length human sequence (M1-V465) suitable for functional and binding studies.
  • N-terminal SUMO and 6xHis tags for enhanced solubility and affinity purification.
  • Lyophilized formulation from Tris-HCl/NaCl with TCEP, Brij-35, and glycerol.
  • High purity (>95%) by reducing SDS-PAGE, minimizing assay interference.
  • Stable frozen; aliquot and store at -20°C or -80°C after reconstitution for long-term use.
  • Validated for antibody binding in immunoassays (reported ED50 ~79 ng/mL).