Learn More
Eurofins Discoverx PRKX, active, 250 µg. (14-677M)
Supplier:  Eurofins Discoverx 14677M
Encompass
N-terminal 6xHis-tagged recombinant, human full length PRKX. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9172