Learn More
Abcam PRL-3 Inhibitor I, 5MG
Catalog No. 502671534
Encompass_Preferred
MW 485.2 Da, Purity =98%. A potent inhibitor of PRL-3, IC₅₀=0.9 μM. Sensitizes PRL-3-expressing cancer cells to chemotherapeutics. Inhibits dephosphorylation of tyrosine-783 of integrin β1 in BGC823 and SW480 cells. Reduces the invasive properties of mouse melanoma B16F10 cells in a cellular model.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1534 Supplier Abcam Supplier No. AB2824215MG
Add to Cart
Add to Cart
MW 485.2 Da, Purity =98%. A potent inhibitor of PRL-3, IC₅₀=0.9 μM. Sensitizes PRL-3-expressing cancer cells to chemotherapeutics. Inhibits dephosphorylation of tyrosine-783 of integrin β1 in BGC823 and SW480 cells. Reduces the invasive properties of mouse melanoma B16F10 cells in a cellular model.
The product is subject to the following: Abcam Restricted Use Statement