Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PRL-3 Inhibitor I | 893449-38-2 | 99.10% | 485.21 | 50 MG
SDP

Catalog No. 5000436704
Encompass_Preferred

PRL-3 Inhibitor I is a potent PRL-3 inhibitor with an IC50 of 0.9 μM, and it reduces invasion in cell-based assays.

  • Potent PRL-3 inhibitor with an IC50 of 0.9 μM.
  • Reduces invasion in cell-based assays.
  • Chemical formula: C17H11Br2NO2S2
  • Molecular weight: 485.21
  • Appearance: Light yellow to green yellow solid.
  • Soluble in DMSO at 41.67 mg/mL.
  • Insoluble in H2O.
  • Solid storage: 4°C, protected from light.
  • Solution storage: -80°C for 6 months or -20°C for 1 month, protected from light.

Catalog No. 50-004-36704 Supplier Medchemexpress LLC Supplier No. HY11247650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PRL-3 Inhibitor I is a potent PRL-3 inhibitor with an IC50 of 0.9 μM, and it reduces invasion in cell-based assays.

  • Potent PRL-3 inhibitor with an IC50 of 0.9 μM.
  • Reduces invasion in cell-based assays.
  • Chemical formula: C17H11Br2NO2S2
  • Molecular weight: 485.21
  • Appearance: Light yellow to green yellow solid.
  • Soluble in DMSO at 41.67 mg/mL.
  • Insoluble in H2O.
  • Solid storage: 4°C, protected from light.
  • Solution storage: -80°C for 6 months or -20°C for 1 month, protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.