Learn More
Cayman Chemical PRL-3 Inhibitor, Purity: ≥95%, CAS: 893449-38-2, 25 mg
SDP
Catalog No. 502966191
Encompass_Preferred
A cell-permeable benzylidene rhodamine that inhibits PRL-3 (IC50 = 900 nM for human PRL-3 in vitro), with minimal activity against other phosphatases; reduces the invasion of mouse melanoma B16F10 cells in a cell-based assay
Catalog No. 50-296-6191 Supplier Cayman Chemical Supplier No. 1871925 MG
A cell-permeable benzylidene rhodamine that inhibits PRL-3 (IC50 = 900 nM for human PRL-3 in vitro), with minimal activity against other phosphatases; reduces the invasion of mouse melanoma B16F10 cells in a cell-based assay