Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 5618MSM3P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: B6.2 + PRLR742. Clonality: Monoclonal. Host: Mouse. Isotype: IgG. Positive Control: Testicular Carcinoma. Cellular Staining: Cell surface, cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Semi-purified human prolactin receptor (B6.2); Recombinant human prolactin receptor (PRLR742). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 70kDa.Chromosome: 5p13.2. Gene Symbol: PRLR. Gene ID: 5618. Ensemble: ENSG00000113494. Uniprot: NP_000940.
Catalog No. 50-188-5084