Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Prlx-93936 (dihydrochloride) | 1094210-96-4 | 10 MG
SDP

Catalog No. 5000234668
Encompass_Preferred

Research-grade HIF-1α inhibitor provided as the dihydrochloride salt for in vitro research. Reported to inhibit HIF-1α and the activated Ras pathway; intended for use in anticancer pathway studies and compound screening. Molecular formula C21H26Cl2N4O2; molecular weight 437.36; CAS 1094210-96-4.

  • Dihydrochloride salt, suitable for laboratory research.
  • Reported HIF-1α inhibitory activity and Ras pathway inhibition.
  • Supplied in small mg package sizes for assay and screening use.
  • Molecular formula C21H26Cl2N4O2; molecular weight 437.36.
  • CAS number 1094210-96-4 for unambiguous identification.

Catalog No. 50-002-34668 Supplier Medchemexpress LLC Supplier No. HY119264A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Research-grade HIF-1α inhibitor provided as the dihydrochloride salt for in vitro research. Reported to inhibit HIF-1α and the activated Ras pathway; intended for use in anticancer pathway studies and compound screening. Molecular formula C21H26Cl2N4O2; molecular weight 437.36; CAS 1094210-96-4.

  • Dihydrochloride salt, suitable for laboratory research.
  • Reported HIF-1α inhibitory activity and Ras pathway inhibition.
  • Supplied in small mg package sizes for assay and screening use.
  • Molecular formula C21H26Cl2N4O2; molecular weight 437.36.
  • CAS number 1094210-96-4 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.