Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Prlx-93936 (dihydrochloride) | 1094210-96-4 | 99.8% | 1 MG
SDP

Catalog No. 5000233164
Encompass_Preferred

PRLX-93936 dihydrochloride is a small-molecule HIF-1α inhibitor with reported anticancer activity that can suppress the activated Ras signaling pathway and is supplied as a solid research reagent for preclinical studies.

  • Inhibits HIF-1α and suppresses Ras pathway signaling.
  • High reported purity (99.8%).
  • Solid form, suitable for storage and formulation.
  • Molecular weight 437.36 g/mol; formula C21H26Cl2N4O2.
  • Recommended storage: 4°C sealed; in solution -80°C (6 months) or -20°C (1 month).
  • Intended for research use only; not for human use.

Catalog No. 50-002-33164 Supplier Medchemexpress LLC Supplier No. HY119264A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PRLX-93936 dihydrochloride is a small-molecule HIF-1α inhibitor with reported anticancer activity that can suppress the activated Ras signaling pathway and is supplied as a solid research reagent for preclinical studies.

  • Inhibits HIF-1α and suppresses Ras pathway signaling.
  • High reported purity (99.8%).
  • Solid form, suitable for storage and formulation.
  • Molecular weight 437.36 g/mol; formula C21H26Cl2N4O2.
  • Recommended storage: 4°C sealed; in solution -80°C (6 months) or -20°C (1 month).
  • Intended for research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.