Learn More
Electron Microscopy Sciences Premium Plus 100-Place Slidebox, Purple
SDP
Catalog No. 501929969
Encompass_Preferred
These slideboxes feature a slide rail design that ensures a secure fit with easy placement and removal of slides. Centrally positioned location numbering grid enables simple slide organization with easy viewing of location numbers. They are made from ABS and Polypropylene, which makes the boxes robust and eliminates potential rusting of metal parts. Hinged lid smoothly opens 180° sitting flat on surfaces and prevents misplacement. The slide lock has easily identified lock and unlock symbols and provides secure storage. A 2 mm zip tie can be used for more secure storage, if desired. Holds 100 26 x 75 mm slides.
Color: Purple
Catalog No. 50-192-9969 Supplier Electron Microscopy Sciences Supplier No. 7149805
These slideboxes feature a slide rail design that ensures a secure fit with easy placement and removal of slides. Centrally positioned location numbering grid enables simple slide organization with easy viewing of location numbers. They are made from ABS and Polypropylene, which makes the boxes robust and eliminates potential rusting of metal parts. Hinged lid smoothly opens 180° sitting flat on surfaces and prevents misplacement. The slide lock has easily identified lock and unlock symbols and provides secure storage. A 2 mm zip tie can be used for more secure storage, if desired. Holds 100 26 x 75 mm slides.
Color: Purple