Learn More
Electron Microscopy Sciences EMS Iris Scissors Premium with Straight Blades
SDP
Catalog No. 501931300
Encompass_Preferred
These Iris scissors are made with sharp tips and are perfect for precise vascular and neurological dissection; ideal for small specimens and animal dissection.
OAL: 114 mm
This item is not returnable. View return policy
Catalog No. 50-193-1300 Supplier Electron Microscopy Sciences Supplier No. 7299816
These Iris scissors are made with sharp tips and are perfect for precise vascular and neurological dissection; ideal for small specimens and animal dissection.
OAL: 114 mm