Learn More
Epigentek Inc Epigenase PRMT Methyltransferase (Type II-Specific) Activity/Inhibition Assay Kit (Colorimetric) (48 assays)
SDP
Supplier:  Epigentek Inc P308848
Encompass_Preferred
Epigenase PRMT Methyltransferase (Type II-Specific) Activity/Inhibition Assay Kit (Colorimetric) is a complete set of optimized buffers and reagents for measuring the activity or inhibition of total type II PRMT using nuclear extracts or purified enzymes such as PRMT5 and PRMT7 from a broad range of species such as mammals, plants, fungi, and bacteria, in a variety of forms including cultured cells and fresh tissues.
Catalog No. 50-109-6328