Learn More
Eurofins Discoverx PRMT1, 50 µg. (EPI053)
Supplier:  Eurofins Discoverx EPI053
Encompass
PRMT1 is purified, N terminal 6xHis tagged recombinant PRMT1 enzyme. This enzyme contains amino acids 211-531. It retains its binding/enzymatic activity and is intended for use in epigenetic assays. This enzyme is provided as 0.54 mg/mL frozen in 50% glycerol. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-200-5266