Learn More
Aviva Systems Biology Corp PRMT2 Antibody - middle region (ARP40202_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40202_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRMT2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PRMT2. Alternate names: HRMT1L1, MGC111373ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-57336