Learn More
Aviva Systems Biology Corp PRMT2 Antibody - N-terminal region (ARP40195_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40195_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRMT2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PRMT2. Alternate names: HRMT1L1, MGC111373ApplicationWB, CHIP. Species reactivity: Human, Rat
Catalog No. 50-014-57332