Learn More
Eurofins Discoverx PRMT3, 50 µg. (EPI054)
Supplier:  Eurofins Discoverx EPI054
Encompass
PRMT3 is purified, N terminal 6xHis tagged recombinant PRMT3 enzyme. This enzyme contains amino acids 843-1208. It retains its binding/enzymatic activity and is intended for use in epigenetic assays. This enzyme is provided as 1 mg/mL frozen in 50% glycerol. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-200-5268