Learn More
Aviva Systems Biology Corp PRMT3 Antibody - middle region (ARP40183_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40183_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRMT3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PRMT3. Alternate names: HRMT1L3ApplicationWB, CHIP. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57326