Learn More
Aviva Systems Biology Corp PRMT5 Antibody - N-terminal region (ARP40185_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40185_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PRMT5. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human PRMT5. Alternate names: JBP1, SKB1, IBP72, SKB1Hs, HRMT1L5ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57327