Learn More
Medchemexpress LLC PRMT5-IN-1 hydrochloride | 99.5% | 455.29 | 1 MG
SDP
Catalog No. 5000436253
Encompass_Preferred
PRMT5 IN-1 hydrochloride (compound 9) is a potent, selective protein arginine methyltransferase 5 (PRMT5) inhibitor. It functions as a hemiaminal and can be converted to aldehydes, reacting with C449 to form covalent adducts under physiological conditions. This compound inhibits cellular sDMA expression and cell proliferation in Granta-519 cells.
  • Potent, selective PRMT5 inhibitor
  • IC50 of 11 nM for PRMT5/MEP50
  • Inhibits cellular sDMA expression with an IC50 of 0.012 μM
  • Inhibits cell proliferation in Granta-519 cells with an IC50 of 0.06 μM
  • Forms covalent adducts with C449
Catalog No. 50-004-36253 Supplier Medchemexpress LLC Supplier No. HY126256A1MG
PRMT5 IN-1 hydrochloride (compound 9) is a potent, selective protein arginine methyltransferase 5 (PRMT5) inhibitor. It functions as a hemiaminal and can be converted to aldehydes, reacting with C449 to form covalent adducts under physiological conditions. This compound inhibits cellular sDMA expression and cell proliferation in Granta-519 cells.
  • Potent, selective PRMT5 inhibitor
  • IC50 of 11 nM for PRMT5/MEP50
  • Inhibits cellular sDMA expression with an IC50 of 0.012 μM
  • Inhibits cell proliferation in Granta-519 cells with an IC50 of 0.06 μM
  • Forms covalent adducts with C449