Learn More
Medchemexpress LLC PRMT5-IN-1 hydrochloride | 99.5% | 455.29 | 10 MG
SDP
Catalog No. 5000436284
Encompass_Preferred
PRMT5 IN-1 hydrochloride (compound 9), a hemiaminal, is a potent, selective protein arginine methyltransferase 5 (PRMT5) inhibitor with an IC50 of 11 nM for PRMT5/MEP50. It can be converted to aldehydes and react with C449 to form covalent adducts under physiological conditions. It is for research use only and not sold to patients.
  • Potent, selective protein arginine methyltransferase 5 (PRMT5) inhibitor
  • IC50 of 11 nM for PRMT5/MEP50
  • Inhibits the PRMT5/MEP50 complex very rapidly with a Kinact of 0.068 min-1 and a good binding affinity (KI=55 nM)
  • Inhibits the expression of cellular sDMA with an IC50 value of 0.012 μM in Granta-519 cells
  • Inhibits cell proliferation in a dose-dependent manner with an IC50 value of 0.06 μM in Granta-519 cells
Catalog No. 50-004-36284 Supplier Medchemexpress LLC Supplier No. HY126256A10MG
PRMT5 IN-1 hydrochloride (compound 9), a hemiaminal, is a potent, selective protein arginine methyltransferase 5 (PRMT5) inhibitor with an IC50 of 11 nM for PRMT5/MEP50. It can be converted to aldehydes and react with C449 to form covalent adducts under physiological conditions. It is for research use only and not sold to patients.
  • Potent, selective protein arginine methyltransferase 5 (PRMT5) inhibitor
  • IC50 of 11 nM for PRMT5/MEP50
  • Inhibits the PRMT5/MEP50 complex very rapidly with a Kinact of 0.068 min-1 and a good binding affinity (KI=55 nM)
  • Inhibits the expression of cellular sDMA with an IC50 value of 0.012 μM in Granta-519 cells
  • Inhibits cell proliferation in a dose-dependent manner with an IC50 value of 0.06 μM in Granta-519 cells