Learn More
Medchemexpress LLC PRMT5-IN-1 hydrochloride | 99.5% | 455.29 | C19H20Cl2N4O5 | 5 MG
SDP
Catalog No. 5000264666
Encompass_Preferred
PRMT5-IN-1 hydrochloride is the hydrochloride salt of a potent, selective inhibitor of protein arginine methyltransferase 5 (PRMT5) for biochemical and cellular research. It exhibits low-nanomolar activity (reported IC50 ≈ 11 nM for PRMT5/MEP50), and the hemiaminal can convert to aldehydes and form covalent adducts under physiological conditions. The material is provided in research-scale quantities and characterized for purity.
Catalog No. 50-002-64666 Supplier Medchemexpress LLC Supplier No. HY126256A5MG
PRMT5-IN-1 hydrochloride is the hydrochloride salt of a potent, selective inhibitor of protein arginine methyltransferase 5 (PRMT5) for biochemical and cellular research. It exhibits low-nanomolar activity (reported IC50 ≈ 11 nM for PRMT5/MEP50), and the hemiaminal can convert to aldehydes and form covalent adducts under physiological conditions. The material is provided in research-scale quantities and characterized for purity.