Learn More
Cayman Chemical PRMT5MEP50 Complexhumnr 20ug
SDP
Catalog No. 502946632
Encompass_Preferred
Source: Active recombinant human N-terminal DYKDDDDK-tagged PRMT5 and N-terminal His-tagged MEP50 expressed in insect cells • Amino acids: 2-2,304 (PRMT5) and 2-2,456 (MEP50) • MW: 73 (PRMT5) and 37.5 (MEP50) kDa
Catalog No. 50-294-6632 Supplier Cayman Chemical Supplier No. 3399220 UG
Source: Active recombinant human N-terminal DYKDDDDK-tagged PRMT5 and N-terminal His-tagged MEP50 expressed in insect cells • Amino acids: 2-2,304 (PRMT5) and 2-2,456 (MEP50) • MW: 73 (PRMT5) and 37.5 (MEP50) kDa