Learn More
Aviva Systems Biology Corp Prmt6 Antibody - C-terminal region (ARP40189_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40189_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Prmt6. It was validated on Western Blot. . Alternate names: Hrmt1l6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-284-858