Learn More
Aviva Systems Biology Corp PRMT8 Antibody - C-terminal region (ARP40193_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40193_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PRMT8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PRMT8. Alternate names: HRMT1L3, HRMT1L4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57331