Learn More
Biomatik Corporation Prn Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11892
Encompass_Preferred
Prn Polyclonal Antibody is a Bordetella pertussis (strain Tohama I / ATCC BAA-589 / NCTC 13251) Polyclonal Antibody to prn. The Prn Polyclonal Antibody has been validated for the following applications: ELISA. Host: Rabbit. Species: Bordetella pertussis (strain Tohama I / ATCC BAA-589 / NCTC 13251). Immunogen: Recombinant Bordetella Pertussis Pertactin Autotransporter Protein (632-910Aa). Applications: ELISA. Target Synonyms: prn. Accession Number: P14283. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6714