Learn More
Medchemexpress LLC HY-112166 10mg Medchemexpress, PRN1008 CAS:1575596-29-0 Purity:>98%
SDP
Catalog No. 501969047
Encompass_Preferred
Medchemexpress, HY-112166 10mg PRN1008 CAS:1575596-29-0 PRN1008 is a reversible covalent, selective and oral active inhibitor of Bruton’s Tyrosine Kinase (BTK), with an IC50 of 1.3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9047 Supplier Medchemexpress LLC Supplier No. HY11216610MG
Medchemexpress, HY-112166 10mg PRN1008 CAS:1575596-29-0 PRN1008 is a reversible covalent, selective and oral active inhibitor of Bruton’s Tyrosine Kinase (BTK), with an IC50 of 1.3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.